Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113541.1 | NP_001107013.1 | 4 | 1345 | ||
XM_011545671.2 | XP_011543973.1 | 4 | 1416 |
Gene Symbol: SPATA31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085452.2 | NP_001078921.2 | 4 | 1378 | ||
XM_011518006.1 | XP_011516308.1 | 4 | 1416 | |||
GenBank mRNA | AK302569.1 | - | 4 | 1378 | ||
BC146855.1 | - | 4 | 1337 | 9056783 |
Gene Symbol: SPATA31A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083124.1 | NP_001076593.1 | 4 | 1345 | ||
XM_011545680.2 | XP_011543982.1 | 3 | 1262 | |||
GenBank mRNA | AL080148.1 | - | 1 | 300 |
Gene Symbol: SPATA31A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145196.1 | NP_001138668.1 | 4 | 1345 | ||
XM_011517871.2 | XP_011516173.1 | 4 | 1352 |
Gene Symbol: SPATA31A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015667.2 | NP_056482.2 | 4 | 1345 | ||
GenBank mRNA | BC040528.1 | - | 4 | 1345 | 4838470 |