Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136329.1 | 3 | 216 | |||
GenBank mRNA | DB034577.1 | - | 3 | 216 |
Gene Symbol: LOC101927429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136328.1 | 3 | 216 | |||
GenBank mRNA | AK310441.1 | - | 3 | 219 |
Gene Symbol: LOC107983967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003062.1 | XP_016858551.1 | 3 | 239 |