Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KMT2C SIRNA Assays ›
Gene Symbol:
KMT2C
LOC107983967
Gene Name:
lysine methyltransferase 2C
histone-lysine N-methyltransferase 2C-like
Gene Aliases:
HALR, MLL3
-
Chromosome Location:
Chr.7: 152134925 - 152436642 on Build GRCh38
Chromosome Location:
Chr.1: 143464779 - 143476935 on Build GRCh38
UniGene ID:
Hs.647120
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s55099
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KMT2C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_170606.2
NP_733751.2
18, 19
3193
XM_005250025.4
XP_005250082.1
19, 20
2999
XM_005250026.3
XP_005250083.1
19, 20
3033
XM_005250027.4
XP_005250084.1
18, 19
3207
XM_005250028.4
XP_005250085.1
18, 19
3207
XM_005250031.4
XP_005250088.1
18, 19
3207
XM_006716077.3
XP_006716140.1
18, 19
3207
XM_006716078.3
XP_006716141.1
18, 19
3207
XM_006716079.3
XP_006716142.1
18, 19
3207
XM_011516450.2
XP_011514752.1
18, 19
3207
XM_011516451.2
XP_011514753.1
17, 18
3087
XM_011516452.2
XP_011514754.1
18, 19
3207
XM_011516453.2
XP_011514755.1
18, 19
3207
XM_011516454.2
XP_011514756.1
13, 14
2202
XM_011516456.2
XP_011514758.1
18, 19
3207
XM_017012480.1
XP_016867969.1
19, 20
3036
XM_017012481.1
XP_016867970.1
19, 20
2996
XM_017012482.1
XP_016867971.1
18, 19
3207
XM_017012483.1
XP_016867972.1
18, 19
3207
XM_017012484.1
XP_016867973.1
18, 19
3174
XM_017012485.1
XP_016867974.1
18, 19
3204
XM_017012486.1
XP_016867975.1
18, 19
3207
XM_017012487.1
XP_016867976.1
18, 19
3175
XM_017012488.1
XP_016867977.1
18, 19
3207
GenBank mRNA
AB040939.2
-
7, 8
1309
AF264750.2
-
4, 5
520
AK025911.1
-
8, 9
1419
AK095628.1
-
11, 12
1915
AY024361.1
-
18, 19
3154
BC151837.1
-
7, 8
1309
Gene Symbol: LOC107983967
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017003062.1
XP_016858551.1
4, 5
396
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008762670.This siRNA also targets the following transcripts in Mouse: NM_001081383, AY036886, AK087368, AY138582, BC037044.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
histone H3-K4 methylation
DNA binding
protein binding
zinc ion binding
transferase activity, transferring acyl groups
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0