Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162995.2 | NP_001156467.1 | 3 | 1219 | ||
NR_028439.1 | 2 | 4021 | ||||
XM_011525118.2 | XP_011523420.1 | 3 | 561 | |||
XM_011525119.2 | XP_011523421.1 | 3 | 469 | |||
XM_017024945.1 | XP_016880434.1 | 4 | 768 | |||
XM_017024946.1 | XP_016880435.1 | 3 | 1600 | |||
XM_017024947.1 | XP_016880436.1 | 3 | 718 | |||
XM_017024948.1 | XP_016880437.1 | 4 | 666 | |||
GenBank mRNA | AF370408.1 | - | 2 | 488 | ||
AK055768.1 | - | 2 | 2182 | |||
AL832643.2 | - | 2 | 4021 | |||
BC146918.1 | - | 2 | 1244 | |||
BC146923.1 | - | 2 | 1244 | |||
BI712658.1 | - | 3 | 582 |