Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SMIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048249.3 | NP_001041714.1 | 3 | 1235 | ||
GenBank mRNA | BC014242.2 | - | 3 | 1086 | ||
BC047489.2 | - | 3 | 1116 | |||
BC066550.1 | - | 3 | 1214 | |||
BC103711.1 | - | 3 | 1190 | |||
BX538015.1 | - | 3 | 1230 | |||
BX648459.1 | - | 3 | 1207 |