Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000818.2 | NP_000809.1 | 7 | 1334 | ||
NM_001134366.1 | NP_001127838.1 | 7 | 1334 | |||
GenBank mRNA | AJ251501.1 | - | 4 | 334 | ||
BC126327.1 | - | 7 | 856 | 8992043 | ||
BC126329.1 | - | 7 | 856 | 8992045 | ||
M74826.1 | - | 7 | 972 | |||
M81882.1 | - | 7 | 883 | |||
X69936.1 | - | 7 | 876 |