Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123376.2 | NP_001116848.1 | 5 | 954 | ||
XM_011540069.2 | XP_011538371.1 | 5 | 3756 | |||
XM_011540070.2 | XP_011538372.1 | 5 | 3702 | |||
XM_011540071.2 | XP_011538373.1 | 6 | 1063 | |||
GenBank mRNA | AB235418.1 | - | 5 | 909 | ||
BC127789.1 | - | 4 | 887 | 40133552 | ||
BC127790.1 | - | 5 | 954 | 40133554 |