Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000818.2 | NP_000809.1 | 5 | 1069 | ||
NM_001134366.1 | NP_001127838.1 | 5 | 1069 | |||
GenBank mRNA | BC126327.1 | - | 5 | 591 | 8992043 | |
BC126329.1 | - | 5 | 591 | 8992045 | ||
M74826.1 | - | 5 | 707 | |||
M81882.1 | - | 5 | 618 | |||
X69936.1 | - | 5 | 611 |