Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034111.1 | 2, 3 | 220 | |||
GenBank mRNA | BC031311.1 | - | 1, 2 | 486 | ||
BX647114.1 | - | 2, 3 | 176 |