Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039770.2 | NP_001034859.2 | 2 | 1457 | ||
NM_001136238.1 | NP_001129710.1 | 2 | 942 | |||
GenBank mRNA | AK126979.1 | - | 2 | 1457 | ||
AK128242.1 | - | 2 | 942 | |||
BC136872.1 | - | 1 | 1383 | 9020863 | ||
BC136873.1 | - | 1 | 1383 | 9020864 |