Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020474.3 | NP_065207.2 | 7, 8 | 1262 | ||
XM_005258239.3 | XP_005258296.1 | 8, 9 | 1330 | |||
XM_017025692.1 | XP_016881181.1 | 8, 9 | 1509 | |||
GenBank mRNA | AK293062.1 | - | 9, 10 | 1404 | ||
BC038440.1 | - | 8, 9 | 1327 | 4478643 | ||
U41514.1 | - | 7, 8 | 1199 | |||
X85018.1 | - | 7, 8 | 1168 |