Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002295.1 | NP_001002295.1 | 6 | 1670 | ||
NM_002051.2 | NP_002042.1 | 6 | 1667 | |||
XM_005252442.2 | XP_005252499.1 | 6 | 1652 | |||
XM_005252443.4 | XP_005252500.1 | 6 | 1577 | |||
GenBank mRNA | AK298213.1 | - | 5 | 897 | ||
BC003070.2 | - | 6 | 1561 | 3504200 | ||
BC006793.1 | - | 6 | 1672 | 2985843 | ||
M69106.1 | - | 5 | 1232 | |||
X55037.1 | - | 5 | 1234 | |||
X55122.1 | - | 5 | 1311 | |||
X58072.1 | - | 5 | 1261 |