Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TUBB SIRNA Assays ›
Gene Symbol:
TUBB
Gene Name:
tubulin beta class I
Gene Aliases:
CDCBM6, CSCSC1, M40, OK/SW-cl.56, TUBB1, TUBB5
Chromosome Location:
Chr.6: 30720201 - 30725426 on Build GRCh38
UniGene ID:
Hs.636480
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s56619
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TUBB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001293212.1
NP_001280141.1
4
1381
NM_001293213.1
NP_001280142.1
5
691
NM_001293214.1
NP_001280143.1
3
1165
NM_001293215.1
NP_001280144.1
4
1284
NM_001293216.1
NP_001280145.1
4
1183
NM_178014.3
NP_821133.1
4
1297
NR_120608.1
4
698
GenBank mRNA
AB062393.1
-
4
1123
AF070561.1
-
4
1118
AF070593.1
-
4
1118
AF070600.1
-
4
1135
AF141349.1
-
4
1135
AK098772.1
-
4
1142
AK297489.1
-
5
445
AK297638.1
-
4
765
AK298889.1
-
4
1187
AK302195.1
-
3
627
AK302314.1
-
4
1250
AK303227.1
-
3
884
AK309684.1
-
2
739
AK316265.1
-
4
1054
AK316316.1
-
4
1179
BC001896.1
-
1
679
BC001938.1
-
4
1122
2820585
BC002347.2
-
4
1127
2964461
BC005838.2
-
4
1122
2820585
BC007605.1
-
4
1127
3350604
BC013374.2
-
4
1118
3946253
BC015889.1
-
1
459
BC019924.1
-
4
1115
4998571
BC020946.1
-
4
1156
4795692
BC021909.1
-
4
1117
3509417
BC029323.1
-
1
480
BC062532.1
-
4
460
BC070326.1
-
4
1133
6698679
BC103746.1
-
4
1140
6503977
DQ457058.1
-
1
553
V00599.1
-
3
970
Back To Top
More Information
Panther Classification:
Molecular Function -
tubulin
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
movement of cell or subcellular component
microtubule-based process
cellular process
cytoskeleton-dependent intracellular transport
natural killer cell mediated cytotoxicity
spindle assembly
cell division
GTPase activity
structural molecule activity
structural constituent of cytoskeleton
protein binding
GTP binding
protein domain specific binding
ubiquitin protein ligase binding
protein complex binding
GTPase activating protein binding
MHC class I protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0