Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DLEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_109974.1 | 3 | 461 | |||
GenBank mRNA | AJ412027.1 | - | LIT1925 BCMS |
1 | 19 | |
AJ412028.1 | - | LIT1924 BCMS |
1 | 19 | ||
AJ412029.1 | - | LIT1923 BCMS |
1 | 19 | ||
AJ412038.1 | - | LIT1907 BCMS |
2 | 406 | ||
AJ412039.1 | - | LIT1906 BCMS |
3 | 459 | ||
AJ412041.1 | - | LIT1904 BCMS |
3 | 325 | ||
AJ412042.1 | - | LIT1903 BCMS |
2 | 210 | ||
AJ412043.1 | - | LIT1902 BCMS |
2 | 210 | ||
AJ412044.1 | - | LIT1898 BCMS |
2 | 46 | ||
AJ412045.1 | - | LIT1897 BCMS |
5 | 368 | ||
AJ412046.1 | - | LIT1894 BCMS |
2 | 116 | ||
AJ412047.1 | - | LIT1893 BCMS |
2 | 235 | ||
AJ412048.1 | - | LIT1892 BCMS |
2 | 199 | ||
AJ412049.1 | - | LIT1891 BCMS |
2 | 131 | ||
AJ412050.1 | - | LIT1890 BCMS |
3 | 564 | ||
AJ412051.1 | - | LIT1889 BCMS |
2 | 485 | ||
AJ412055.1 | - | LIT1885 BCMS |
3 | 459 | ||
AJ412057.1 | - | LIT1876 BCMS |
3 | 459 | ||
AJ412059.1 | - | LIT1874 BCMS |
3 | 459 | ||
AJ412061.1 | - | LIT1872 BCMS |
3 | 459 | ||
AJ412062.1 | - | LIT1871 BCMS |
3 | 459 | ||
AJ412063.1 | - | LIT1870 BCMS |
1 | 19 |