Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114091.2 | NP_001107563.1 | 3 | 360 | ||
NM_001256.4 | NP_001247.3 | 3 | 360 | |||
NM_001293089.1 | NP_001280018.1 | 3 | 360 | |||
XM_011525546.2 | XP_011523848.1 | 3 | 347 | |||
XM_011525547.2 | XP_011523849.1 | 3 | 347 | |||
XM_017025481.1 | XP_016880970.1 | 3 | 347 | |||
XM_017025482.1 | XP_016880971.1 | 3 | 347 | |||
XM_017025483.1 | XP_016880972.1 | 3 | 347 | |||
XM_017025488.1 | XP_016880977.1 | 3 | 327 | |||
GenBank mRNA | AK299342.1 | - | 3 | 312 | ||
AU135593.1 | - | 3 | 348 | |||
BC011656.2 | - | 3 | 224 | 4301175 | ||
BI548077.1 | - | 352 | ||||
DQ786193.1 | - | 3 | 272 | |||
HY023336.1 | - | 3 | 361 | |||
S78234.1 | - | 3 | 320 | |||
U00001.1 | - | 3 | 278 |