Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005260.4 | NP_005251.1 | 1 | 67 | ||
XM_011543308.2 | XP_011541610.1 | 2 | 2038 | |||
GenBank mRNA | BC096228.3 | - | 1 | 67 | 40005256 | |
BC096229.3 | - | 1 | 67 | 40005257 | ||
BC096230.3 | - | 1 | 67 | 40005258 | ||
BC096231.1 | - | 1 | 67 | 40005259 | ||
CA488820.1 | - | 1 | 801 |