Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146282.1 | NP_001139754.1 | 7 | 787 | ||
NM_003136.3 | NP_003127.1 | 8 | 879 | |||
XM_005268024.2 | XP_005268081.1 | 8 | 1132 | |||
XM_011537106.1 | XP_011535408.1 | 8 | 760 | |||
XM_017021615.1 | XP_016877104.1 | 7 | 719 | |||
GenBank mRNA | AK293319.1 | - | 8 | 671 | ||
AK300824.1 | - | 7 | 660 | |||
AK312853.1 | - | 8 | 824 | |||
BC000652.2 | - | 8 | 746 | 3343314 | ||
BC003389.1 | - | 8 | 752 | 3446599 | ||
U51920.1 | - | 8 | 599 |