Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146282.1 | NP_001139754.1 | 9 | 1064 | ||
NM_003136.3 | NP_003127.1 | 10 | 1156 | |||
XM_005268024.2 | XP_005268081.1 | 10 | 1409 | |||
XM_011537106.1 | XP_011535408.1 | 10 | 1037 | |||
XM_017021615.1 | XP_016877104.1 | 9 | 996 | |||
GenBank mRNA | AK293319.1 | - | 10 | 948 | ||
AK300824.1 | - | 9 | 937 | |||
BC000652.2 | - | 10 | 1023 | 3343314 | ||
BC003389.1 | - | 10 | 1029 | 3446599 | ||
U51920.1 | - | 10 | 876 |