Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288833.1 | NP_001275762.1 | 10, 11 | 1676 | ||
NM_013421.2 | NP_038265.2 | 11, 12 | 1604 | |||
NM_013430.2 | NP_038347.2 | 10, 11 | 1364 | |||
GenBank mRNA | AL832738.1 | - | 1580 | |||
BC025927.1 | - | 8, 9 | 1244 | 4548861 | ||
CR456494.1 | - | 7, 8 | 954 | |||
DQ000492.2 | - | 1, 2 | 59 | |||
J04131.1 | - | 11, 12 | 1546 | |||
J05235.1 | - | 9, 10 | 1621 | |||
M24087.1 | - | 6, 7 | 879 | |||
M24903.1 | - | 10, 11 | 1364 | |||
X60069.1 | - | 8, 9 | 1235 |
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261890.4 | XP_005261947.1 | 3, 4 | 430 | ||
XM_006724392.3 | XP_006724455.1 | 7, 8 | 1138 | |||
XM_006724395.3 | XP_006724458.1 | 4, 5 | 590 | |||
XM_017029179.1 | XP_016884668.1 | 8, 9 | 1819 | |||
XM_017029180.1 | XP_016884669.1 | 7, 8 | 1277 | |||
XM_017029181.1 | XP_016884670.1 | 7, 8 | 1715 | |||
XM_017029182.1 | XP_016884671.1 | 8, 9 | 1819 | |||
XM_017029183.1 | XP_016884672.1 | 3, 4 | 406 | |||
XM_017029185.1 | XP_016884674.1 | 9, 10 | 2659 | |||
XM_017029186.1 | XP_016884675.1 | 12, 13 | 2282 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 7, 8 | 1071 | |||
GenBank mRNA | BC143607.1 | - | 4, 5 | 563 |
Gene Symbol: LOC102724197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724921.3 | XP_006724984.1 | 7, 8 | 1581 | ||
XM_006724924.3 | XP_006724987.1 | 6, 7 | 1423 | |||
XM_006724928.3 | XP_006724991.1 | 2, 3 | 619 | |||
XM_011546256.2 | XP_011544558.1 | 8, 9 | 1715 | |||
XM_017030133.1 | XP_016885622.1 | 8, 9 | 1146 | |||
XM_017030134.1 | XP_016885623.1 | 8, 9 | 1149 | |||
XM_017030135.1 | XP_016885624.1 | 8, 9 | 1574 | |||
XM_017030136.1 | XP_016885625.1 | 9, 10 | 1616 | |||
XM_017030137.1 | XP_016885626.1 | 9, 10 | 1613 | |||
XM_017030138.1 | XP_016885627.1 | 9, 10 | 1136 | |||
XM_017030139.1 | XP_016885628.1 | 9, 10 | 1139 | |||
XM_017030140.1 | XP_016885629.1 | 10, 11 | 1606 | |||
XM_017030141.1 | XP_016885630.1 | 10, 11 | 1603 | |||
XM_017030142.1 | XP_016885631.1 | 8, 9 | 1447 | |||
XM_017030143.1 | XP_016885632.1 | 7, 8 | 1582 | |||
XM_017030144.1 | XP_016885633.1 | 3, 4 | 510 | |||
XM_017030145.1 | XP_016885634.1 | 4, 5 | 1143 | |||
XM_017030146.1 | XP_016885635.1 | 7, 8 | 1598 |
Gene Symbol: LOC102724823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030162.1 | XP_016885651.1 | 8, 9 | 1615 |