Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4DIP SIRNA Assays ›
Gene Symbol:
PDE4DIP
LOC100996724
Gene Name:
phosphodiesterase 4D interacting protein
phosphodiesterase 4D interacting protein-like
Gene Aliases:
CMYA2, MMGL
-
Chromosome Location:
Chr.1: 148808167 - 149051277 on Build GRCh38
Chromosome Location:
Chr.1: 120471449 - 120701470 on Build GRCh38
UniGene ID:
Hs.584841
Hs.731111
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s57385
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4DIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002811.2
NP_001002811.2
3
1944
NM_001002812.2
NP_001002812.2
7
1100
NM_001198832.2
NP_001185761.2
10
1382
NM_001198834.3
NP_001185763.3
7
1100
NM_014644.5
NP_055459.5
7
1100
XM_005272981.3
XP_005273038.2
3
1727
XM_005277442.4
XP_005277499.2
3
1727
XM_005277443.3
XP_005277500.1
3
1727
XM_006711642.2
XP_006711705.1
3
1727
XM_006711643.2
XP_006711706.1
3
1727
XM_006711644.2
XP_006711707.1
3
1727
XM_006711646.1
XP_006711709.1
10
1389
XM_006711648.2
XP_006711711.1
3
1727
XM_006711650.3
XP_006711713.1
8
874
XM_006711651.1
XP_006711714.1
7
992
XM_006711652.1
XP_006711715.1
7
992
XM_011510172.1
XP_011508474.1
10
1547
XM_011510173.1
XP_011508475.1
10
1547
XM_011510174.1
XP_011508476.1
3
1727
XM_011510175.1
XP_011508477.1
10
1547
XM_011510176.1
XP_011508478.1
10
1547
XM_011510179.1
XP_011508481.1
7
992
XM_017002875.1
XP_016858364.1
3
1727
XM_017002876.1
XP_016858365.1
3
1727
XM_017002877.1
XP_016858366.1
3
1727
XM_017002878.1
XP_016858367.1
10
1547
XM_017002879.1
XP_016858368.1
3
1727
XM_017002880.1
XP_016858369.1
10
1547
XM_017002881.1
XP_016858370.1
3
1727
XM_017002882.1
XP_016858371.1
3
1727
XM_017002883.1
XP_016858372.1
3
1727
XM_017002884.1
XP_016858373.1
3
1727
XM_017002885.1
XP_016858374.1
3
1727
XM_017002886.1
XP_016858375.1
3
1727
XM_017002887.1
XP_016858376.1
11
1824
XM_017002888.1
XP_016858377.1
11
1797
XM_017002889.1
XP_016858378.1
11
1469
XM_017002890.1
XP_016858379.1
3
1727
XM_017002891.1
XP_016858380.1
9
1628
XM_017002892.1
XP_016858381.1
9
1259
XM_017002893.1
XP_016858382.1
10
1747
XM_017002894.1
XP_016858383.1
10
1392
XM_017002895.1
XP_016858384.1
7
710
XM_017002896.1
XP_016858385.1
3
1727
XM_017002897.1
XP_016858386.1
3
1727
XM_017002898.1
XP_016858387.1
10
1389
XM_017002899.1
XP_016858388.1
10
1389
XM_017002900.1
XP_016858389.1
3
1727
XM_017002901.1
XP_016858390.1
3
1727
XM_017002902.1
XP_016858391.1
10
1389
XM_017002903.1
XP_016858392.1
10
1389
GenBank mRNA
AB007923.2
-
10
1261
AB007946.1
-
3
1944
AL831815.2
-
7
943
AL832024.2
-
7
908
AL833273.1
-
7
988
BC004860.2
-
3
1611
BC078660.1
-
3
1450
BC132717.1
-
3
1547
BC152439.1
-
3
1944
BX647660.1
-
7
1028
HQ333476.1
-
3
1357
Gene Symbol: LOC100996724
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017003064.1
XP_016858553.1
10
1543
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein complex assembly
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0