Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FAM228A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040710.2 | NP_001035800.1 | 4, 5 | 404 | ||
GenBank mRNA | BC050462.1 | - | 4, 5 | 344 | 5744344 | |
HY022970.1 | - | 4, 5 | 405 |