Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005720.3 | NP_005711.1 | 3 | 218 | ||
XM_006715825.1 | XP_006715888.1 | 3 | 229 | |||
XM_006715826.1 | XP_006715889.1 | 3 | 213 | |||
GenBank mRNA | AF006084.1 | - | 3 | 178 | ||
AK314818.1 | - | 3 | 163 | |||
AU143525.1 | - | 3 | 218 | |||
AY007111.1 | - | 1 | 48 | |||
BC002562.2 | - | 3 | 151 | 3138319 | ||
BC002988.1 | - | 3 | 187 | 2821335 | ||
BC007555.1 | - | 3 | 149 | 2967684 |