Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100289279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545672.2 | XP_011543974.1 | 3 | 604 | ||
XM_011545673.2 | XP_011543975.1 | 3 | 604 | |||
XM_011545674.2 | XP_011543976.1 | 3 | 604 | |||
XM_017015380.1 | XP_016870869.1 | 3 | 604 | |||
GenBank mRNA | DA717841.1 | - | 3 | 269 |
Gene Symbol: CNTNAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033655.3 | NP_387504.2 | 3 | 467 | ||
GenBank mRNA | AB051501.2 | - | 3 | 374 | ||
AF333769.2 | - | 3 | 335 | |||
AK056833.1 | - | 3 | 467 | |||
AK294281.1 | - | 3 | 259 | |||
AU130948.1 | - | 4 | 573 | |||
BC132737.1 | - | 3 | 372 | 40146759 | ||
BC144130.1 | - | 3 | 372 |
Gene Symbol: CNTNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201380.2 | NP_001188309.2 | 3 | 638 |
Gene Symbol: CNTNAP3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111893.2 | 3 | 327 | |||
GenBank mRNA | AK024257.1 | - | 4 | 603 | ||
AK054645.1 | - | 3 | 491 | |||
DA051083.1 | - | 3 | 381 |