Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031297.4 | NP_112587.2 | 1 | 328 | ||
GenBank mRNA | AL136540.1 | - | 11 | |||
DN995061.1 | - | 1 | 91 |