Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025222.3 | NP_079498.2 | 2, 3 | 539 | ||
GenBank mRNA | AK291380.1 | - | 2, 3 | 295 | ||
AY358264.1 | - | 2, 3 | 274 | |||
BX648628.1 | - | 2, 3 | 281 |