Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166425.1 | NP_001159897.1 | 3 | 281 | ||
NM_001282617.1 | NP_001269546.1 | 3 | 322 | |||
NM_001282618.1 | NP_001269547.1 | 2 | 353 | |||
NM_001282619.1 | NP_001269548.1 | 4 | 729 | |||
NM_001282620.1 | NP_001269549.1 | 3 | 605 | |||
NM_002070.3 | NP_002061.1 | 3 | 396 | |||
GenBank mRNA | AF493906.1 | - | 3 | 217 | ||
AK055574.1 | - | 2 | 306 | |||
AK096299.1 | - | 3 | 498 | |||
AK096595.1 | - | 3 | 1000 | |||
AK126708.1 | - | 3 | 2033 | |||
AK131062.1 | - | 1 | 1044 | |||
AK302330.1 | - | 3 | 250 | |||
AK304473.1 | - | 3 | 161 | |||
AY677118.1 | - | 3 | 217 | |||
BC012138.1 | - | 3 | 377 | 4562585 | ||
BC014627.2 | - | 3 | 334 | 2963802 | ||
BC016995.1 | - | 4 | 680 | 4403631 | ||
CR456783.1 | - | 3 | 217 | |||
DB044421.1 | - | 3 | 281 | |||
HY037331.1 | - | 3 | 396 | |||
J03004.1 | - | 3 | 294 | |||
X04828.1 | - | 3 | 340 |