Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182620.3 | NP_872426.1 | 3 | 413 | ||
XM_011524745.2 | XP_011523047.1 | 3 | 227 | |||
GenBank mRNA | AK056473.1 | - | 3 | 165 | ||
BC017873.1 | - | 3 | 174 | 4685972 | ||
BC106003.1 | - | 3 | 138 | 6143473 |