Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143909.1 | NP_001137381.1 | 1 | 1639 | ||
GenBank mRNA | BC157875.1 | - | 1 | 1640 | 9057093 | |
BC171885.1 | - | 1 | 1641 | 9054539 |