Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000406.2 | NP_000397.1 | 1 | 1968 | ||
NM_001012763.1 | NP_001012781.1 | 1 | 1968 | |||
GenBank mRNA | AY392011.1 | - | 1 | 217 | ||
BC113546.1 | - | 1 | 243 | 8322598 | ||
L03380.1 | - | 1 | 272 | |||
L07949.1 | - | 1 | 241 | |||
S60587.1 | - | 1 | 241 | |||
S77472.1 | - | 1 | 242 | |||
Z81148.1 | - | 1 | 217 |
Set Membership: |
![]() |