Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001740.3 | NP_001001740.1 | 18, 19 | 2373 | ||
NM_001286644.1 | NP_001273573.1 | 17, 18 | 2248 | |||
NM_022457.6 | NP_071902.2 | 19, 20 | 2445 | |||
XM_005245447.2 | XP_005245504.1 | 19, 20 | 2468 | |||
XM_005245448.2 | XP_005245505.1 | 18, 19 | 2420 | |||
XM_006711487.2 | XP_006711550.1 | 18, 19 | 2411 | |||
XM_011509881.1 | XP_011508183.1 | 20, 21 | 2534 | |||
XM_017002076.1 | XP_016857565.1 | 17, 18 | 1632 | |||
GenBank mRNA | AF508940.1 | - | 19, 20 | 2179 | ||
AF527539.1 | - | 19, 20 | 2442 | |||
AK001278.1 | - | 17, 18 | 1683 | |||
AK314750.1 | - | 19, 20 | 2377 | |||
AY509921.1 | - | 18, 19 | 2107 | |||
BC039723.1 | - | 17, 18 | 1695 | |||
BC094728.1 | - | 19, 20 | 2433 | 5271629 | ||
BX537932.1 | - | 7, 8 | 1790 | |||
KJ534928.1 | - | 2179 | ||||
KJ534929.1 | - | 1984 | ||||
KJ534930.1 | - | 2119 | ||||
KJ534931.1 | - | 2110 | ||||
KJ534932.1 | - | 2167 | ||||
KJ534933.1 | - | 1678 | ||||
KJ534992.1 | - | 680 | ||||
KJ535061.1 | - | 1884 | ||||
KJ535076.1 | - | 2081 |