Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000175.3 | NP_000166.2 | 15, 16 | 1541 | ||
NM_001184722.1 | NP_001171651.1 | 15, 16 | 1604 | |||
NM_001289789.1 | NP_001276718.1 | 16, 17 | 1688 | |||
NM_001289790.1 | NP_001276719.1 | 14, 15 | 1457 | |||
XM_005258764.2 | XP_005258821.1 | 16, 17 | 1507 | |||
XM_006723148.2 | XP_006723211.1 | 16, 17 | 1458 | |||
XM_011526754.2 | XP_011525056.1 | 17, 18 | 1923 | |||
GenBank mRNA | AK025421.1 | - | 4, 5 | 1139 | ||
AK129884.1 | - | 16, 17 | 1472 | |||
AK130102.1 | - | 14, 15 | 1236 | |||
AK293446.1 | - | 14, 15 | 1333 | |||
AK294396.1 | - | 15, 16 | 1552 | |||
AK301103.1 | - | 15, 16 | 1345 | |||
BC004982.1 | - | 15, 16 | 1409 | 2906270 | ||
K03515.1 | - | 15, 16 | 1416 | |||
X16539.1 | - | 15, 16 | 1407 |