Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316972.1 | NP_001303901.1 | 6 | 684 | ||
NM_001316994.1 | NP_001303923.1 | 6 | 436 | |||
NR_024330.1 | 5 | 1425 | ||||
XM_017026299.1 | XP_016881788.1 | 6 | 2207 | |||
XM_017026300.1 | XP_016881789.1 | 6 | 567 | |||
XM_017026301.1 | XP_016881790.1 | 6 | 633 | |||
GenBank mRNA | AK024362.1 | - | 184 | |||
AY358799.1 | - | 5 | 1456 | |||
KF751883.1 | - | 6 | 525 |