Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TP53 SIRNA Assays ›
Gene Symbol:
TP53
Gene Name:
tumor protein p53
Gene Aliases:
BCC7, LFS1, P53, TRP53
Chromosome Location:
Chr.17: 7668402 - 7687550 on Build GRCh38
UniGene ID:
Hs.437460
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s606
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TP53
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000546.5
NP_000537.3
6
806
NM_001126112.2
NP_001119584.1
6
803
NM_001126113.2
NP_001119585.1
6
806
NM_001126114.2
NP_001119586.1
6
806
NM_001126115.1
NP_001119587.1
2
486
NM_001126116.1
NP_001119588.1
2
486
NM_001126117.1
NP_001119589.1
2
486
NM_001126118.1
NP_001119590.1
5
923
NM_001276695.1
NP_001263624.1
6
806
NM_001276696.1
NP_001263625.1
6
806
NM_001276697.1
NP_001263626.1
2
486
NM_001276698.1
NP_001263627.1
2
486
NM_001276699.1
NP_001263628.1
2
486
NM_001276760.1
NP_001263689.1
6
806
NM_001276761.1
NP_001263690.1
6
803
GenBank mRNA
AB082923.1
-
6
667
AF307851.1
-
6
739
AK225838.1
-
6
744
AK297462.1
-
7
671
AK297927.1
-
7
716
AK303277.1
-
5
862
AK312568.1
-
6
748
AM076970.1
-
5
571
AM076971.1
-
5
604
AM076972.1
-
5
604
AY429684.1
-
5
604
BC003596.1
-
6
724
3544714
BT019622.1
-
5
604
DQ186648.1
-
6
736
DQ186649.1
-
6
736
DQ186650.1
-
2
482
DQ186651.1
-
2
482
DQ186652.1
-
2
482
DQ191317.1
-
6
793
DQ263704.1
-
5
604
DQ286964.1
-
6
738
DQ648884.1
-
5
404
DQ648885.1
-
3
280
EF101867.1
-
5
571
EF101868.1
-
5
604
EF101869.1
-
5
604
JN214348.1
-
5
649
JN601513.1
-
5
604
JN601514.1
-
5
604
JN601515.1
-
5
604
JN601516.1
-
6
578
JN900492.1
-
5
912
JQ026207.1
-
5
594
JQ026208.1
-
5
599
JQ026209.1
-
5
604
JQ026210.1
-
5
604
JQ694049.1
-
5
627
JQ694050.1
-
5
627
JQ694051.1
-
5
627
KC820708.1
-
2
99
KC820709.1
-
2
99
KC820710.1
-
2
99
KC820711.1
-
2
99
KC820712.1
-
2
99
KC820713.1
-
2
99
KC820714.1
-
2
99
KC820715.1
-
2
99
KC820716.1
-
2
99
KC820717.1
-
2
99
KC820718.1
-
2
99
KC820719.1
-
2
99
KC820720.1
-
2
99
KC820721.1
-
2
99
KC820722.1
-
2
99
KC820723.1
-
2
99
KC820724.1
-
2
99
KC820725.1
-
2
99
KC820726.1
-
2
99
KC820727.1
-
2
99
KC820728.1
-
2
99
KC820729.1
-
2
99
KC820730.1
-
2
99
KC820731.1
-
2
99
KC820732.1
-
2
99
KC820733.1
-
2
99
KC820734.1
-
2
99
KC820735.1
-
2
99
KC820736.1
-
2
99
KC820737.1
-
2
99
KC820738.1
-
2
99
KC820739.1
-
2
99
KC820740.1
-
2
99
KC820741.1
-
2
99
KC820742.1
-
2
99
KC820743.1
-
2
99
KC820744.1
-
2
99
KC820745.1
-
2
99
KC820746.1
-
2
99
KC820747.1
-
2
99
KC820748.1
-
2
99
KC820749.1
-
2
99
KC820750.1
-
2
99
KC820751.1
-
2
99
KC820752.1
-
2
99
KC820753.1
-
2
99
KC820754.1
-
2
99
KC820755.1
-
2
99
KC820756.1
-
2
99
KC820757.1
-
2
99
KC820758.1
-
2
99
KC820760.1
-
2
99
KC820761.1
-
2
99
KC820762.1
-
2
99
KC820763.1
-
2
99
KC820764.1
-
2
99
KC820765.1
-
2
99
KC820766.1
-
2
99
KC820767.1
-
2
99
KC820768.1
-
2
99
KC820769.1
-
2
99
KC820770.1
-
2
99
KC820771.1
-
2
99
KC820772.1
-
2
99
KC820773.1
-
2
99
KC820774.1
-
2
99
KC820775.1
-
2
99
KC820776.1
-
2
99
KC820777.1
-
2
99
KC820778.1
-
2
99
KC820779.1
-
2
99
KC820780.1
-
2
99
KC820781.1
-
2
99
KC820782.1
-
2
99
KC820783.1
-
2
99
KC820784.1
-
2
99
KC820785.1
-
2
99
KC820786.1
-
2
99
M14694.1
-
6
729
M14695.1
-
6
725
X01405.1
-
3
305
X02469.1
-
6
739
X60010.1
-
5
603
X60011.1
-
5
604
X60012.1
-
5
604
X60013.1
-
5
604
X60014.1
-
5
604
X60015.1
-
5
604
X60016.1
-
5
604
X60017.1
-
5
604
X60018.1
-
5
604
X60019.1
-
5
604
X60020.1
-
5
604
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_003749209.
Matching Taqman Gene Expression Assay:
Hs00153340_m1
Panther Classification:
Molecular Function -
P53-like transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA strand renaturation
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
ER overload response
cell cycle arrest
Ras protein signal transduction
multicellular organism development
cell aging
protein localization
cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
response to X-ray
response to gamma radiation
positive regulation of gene expression
viral process
protein sumoylation
cell differentiation
negative regulation of cell growth
DNA damage response, signal transduction by p53 class mediator
positive regulation of histone deacetylation
chromatin assembly
mitotic G1 DNA damage checkpoint
positive regulation of protein oligomerization
cellular response to UV
cellular response to drug
cellular response to glucose starvation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
entrainment of circadian clock by photoperiod
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to antibiotic
positive regulation of protein export from nucleus
regulation of mitochondrial membrane permeability
negative regulation of fibroblast proliferation
circadian behavior
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of helicase activity
protein tetramerization
negative regulation of telomerase activity
positive regulation of thymocyte apoptotic process
positive regulation of cell cycle arrest
cellular response to hypoxia
cellular response to ionizing radiation
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
replicative senescence
oxidative stress-induced premature senescence
intrinsic apoptotic signaling pathway
oligodendrocyte apoptotic process
positive regulation of execution phase of apoptosis
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
regulation of cell cycle G2/M phase transition
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
positive regulation of reactive oxygen species metabolic process
positive regulation of intrinsic apoptotic signaling pathway
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protease binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
copper ion binding
protein binding
ATP binding
transcription factor binding
zinc ion binding
enzyme binding
protein kinase binding
protein phosphatase binding
receptor tyrosine kinase binding
ubiquitin protein ligase binding
histone acetyltransferase binding
identical protein binding
sequence-specific DNA binding
protein self-association
transcription regulatory region DNA binding
protein heterodimerization activity
protein N-terminus binding
chaperone binding
protein phosphatase 2A binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0