Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017572.3 | NP_060042.2 | 14 | 1716 | ||
NM_199054.2 | NP_951009.1 | 14 | 3743 | |||
GenBank mRNA | AB209074.1 | - | 4 | 4053 | ||
AF125532.1 | - | 13 | 1492 | |||
AF237776.1 | - | 13 | 1507 | |||
AK024347.1 | - | 1 | 1942 | |||
AK092536.1 | - | 4 | 2645 | |||
AL137565.1 | - | 10 | 1178 | |||
AL137615.1 | - | 1 | 1427 |