Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ERBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005862.2 | NP_001005862.1 | 6 | 746 | ||
NM_001289936.1 | NP_001276865.1 | 7 | 797 | |||
NM_001289937.1 | NP_001276866.1 | 3 | 521 | |||
NM_001289938.1 | NP_001276867.1 | 6 | 746 | |||
NM_004448.3 | NP_004439.2 | 3 | 521 | |||
NR_110535.1 | 3 | 584 | ||||
GenBank mRNA | AF177761.2 | - | 3 | 272 | ||
AK131568.1 | - | 1 | 952 | |||
AK295195.1 | - | 7 | 797 | |||
AK296908.1 | - | 3 | 336 | |||
AK313683.1 | - | 3 | 475 | |||
BC080193.1 | - | 3 | 482 | |||
BC110392.1 | - | 6 | 711 | |||
BC167147.1 | - | 3 | 263 | 100013444 | ||
BI755753.1 | - | 6 | 711 | |||
DC346286.1 | - | 3 | 521 | |||
KJ534831.1 | - | 3 | 260 | |||
KJ534832.1 | - | 3 | 260 | |||
KJ534964.1 | - | 3 | 260 | |||
M11730.1 | - | 3 | 410 | |||
X03363.1 | - | 3 | 434 |