Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRB10 SIRNA Assays ›
Gene Symbol:
GRB10
Gene Name:
growth factor receptor bound protein 10
Gene Aliases:
GRB-IR, Grb-10, IRBP, MEG1, RSS
Chromosome Location:
Chr.7: 50590063 - 50793462 on Build GRCh38
UniGene ID:
Hs.164060
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6126
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRB10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001549.2
NP_001001549.1
10
1100
NM_001001550.2
NP_001001550.1
11
1318
NM_001001555.2
NP_001001555.1
12
1565
NM_005311.4
NP_005302.3
11
1238
XM_011515304.1
XP_011513606.1
14
1677
XM_011515312.2
XP_011513614.1
13
1965
XM_011515316.1
XP_011513618.1
12
1510
XM_011515323.2
XP_011513625.1
9
3014
XM_017012029.1
XP_016867518.1
13
1897
XM_017012030.1
XP_016867519.1
12
1831
XM_017012031.1
XP_016867520.1
12
1762
XM_017012032.1
XP_016867521.1
18
2274
XM_017012033.1
XP_016867522.1
15
1812
XM_017012034.1
XP_016867523.1
13
1413
XM_017012035.1
XP_016867524.1
19
3323
XM_017012036.1
XP_016867525.1
20
3108
XM_017012037.1
XP_016867526.1
20
2945
XM_017012038.1
XP_016867527.1
19
2895
XM_017012039.1
XP_016867528.1
16
2142
XM_017012040.1
XP_016867529.1
16
2678
XM_017012041.1
XP_016867530.1
18
2799
XM_017012042.1
XP_016867531.1
14
1719
XM_017012043.1
XP_016867532.1
15
1909
XM_017012044.1
XP_016867533.1
11
1508
XM_017012045.1
XP_016867534.1
11
1172
XM_017012046.1
XP_016867535.1
15
2524
XM_017012047.1
XP_016867536.1
20
3507
XM_017012048.1
XP_016867537.1
19
3391
XM_017012049.1
XP_016867538.1
20
3013
XM_017012050.1
XP_016867539.1
19
2963
XM_017012051.1
XP_016867540.1
18
2867
XM_017012052.1
XP_016867541.1
18
3207
XM_017012053.1
XP_016867542.1
19
2829
XM_017012054.1
XP_016867543.1
18
2779
XM_017012055.1
XP_016867544.1
15
2562
XM_017012056.1
XP_016867545.1
14
3394
XM_017012057.1
XP_016867546.1
17
2683
XM_017012058.1
XP_016867547.1
13
1554
XM_017012059.1
XP_016867548.1
13
1676
XM_017012060.1
XP_016867549.1
21
3640
XM_017012061.1
XP_016867550.1
12
1506
XM_017012062.1
XP_016867551.1
11
1267
XM_017012063.1
XP_016867552.1
19
3372
XM_017012064.1
XP_016867553.1
18
3256
XM_017012065.1
XP_016867554.1
16
2708
XM_017012066.1
XP_016867555.1
14
3067
XM_017012067.1
XP_016867556.1
14
1570
XM_017012068.1
XP_016867557.1
14
2436
GenBank mRNA
AB000731.1
-
11
1213
AF000017.1
-
11
1207
AF001534.1
-
11
1238
AJ271366.1
-
14
1677
AK289511.1
-
13
1532
BC024285.1
-
12
1342
4590330
BX649028.1
-
11
1298
D86962.1
-
13
1970
U34355.1
-
10
1100
U66065.1
-
11
1318
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of Wnt signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
negative regulation of phosphorylation
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0