Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGTA1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003191.1 | 7 | 1641 | |||
NR_045211.1 | 6 | 1372 | ||||
GenBank mRNA | AF305838.1 | - | 5 | 985 | ||
AF378122.1 | - | LIT2094 alpha1,3GTP2 |
7 | 1371 | ||
BC045756.1 | - | 6 | 1212 |