Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGTA1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003191.1 | 5, 6 | 841 | |||
NR_045211.1 | 4, 5 | 572 | ||||
GenBank mRNA | AF305838.1 | - | 3, 4 | 185 | ||
AF378121.1 | - | LIT2093 alpha1,3GTP1 |
5, 6 | 571 | ||
AF378122.1 | - | LIT2094 alpha1,3GTP2 |
5, 6 | 571 | ||
AF378123.1 | - | LIT2095 alpha1,3GTP3 |
5, 6 | 571 | ||
BC045756.1 | - | 4, 5 | 412 | |||
DT220151.1 | - | 4, 5 | 364 |