Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002087.3 | NP_002078.1 | 12 | 1717 | ||
XM_005257253.1 | XP_005257310.1 | 12 | 1762 | |||
GenBank mRNA | AF055008.1 | - | 12 | 1523 | ||
AK000607.1 | - | 12 | 1501 | |||
AK023348.1 | - | 9 | 1005 | |||
AK222522.1 | - | 12 | 1501 | |||
AK296090.1 | - | 12 | 1455 | |||
AK303830.1 | - | 9 | 1027 | |||
AY124489.1 | - | 11 | 1479 | |||
BC010577.2 | - | 11 | 1471 | 3457813 | ||
M75161.1 | - | 11 | 1473 | |||
X62320.1 | - | 12 | 1501 |