Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PSAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085382.1 | NP_001078851.1 | 1 | 1085 | ||
GenBank mRNA | AK097698.1 | - | 1 | 368 | ||
BC034552.1 | - | 1 | 1000 | |||
BC068579.1 | - | 1 | 1059 | |||
DQ991252.1 | - | 1 | 1085 |