Search Thermo Fisher Scientific
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007390.3 | NP_031416.3 | 6 | 515 | ||
GenBank mRNA | AF225980.1 | - | 6 | 505 | ||
AF325348.1 | - | 5 | 312 | |||
AK157809.1 | - | 6 | 512 | |||
AY574266.1 | - | 6 | 465 | |||
AY851289.1 | - | 338 | ||||
L37663.1 | - | 6 | 515 |