Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Agt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007428.3 | NP_031454.3 | 2 | 389 | ||
GenBank mRNA | AK005354.1 | - | 2 | 389 | ||
AK018763.1 | - | 2 | 387 | |||
AK139179.1 | - | 2 | 388 | |||
BC019496.1 | - | 1 | 365 | 4211501 | ||
BC028877.1 | - | 1 | 355 | 4194190 | ||
BC054387.1 | - | 374 | 6477694 |