Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175086.3 | NP_780295.2 | 2 | 1166 | ||
GenBank mRNA | AK087228.1 | - | 2 | 1166 | ||
BC109152.1 | - | 2 | 1114 | |||
BC109153.1 | - | 2 | 1113 | 40046982 |
Set Membership: |
![]() |