Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301281.1 | NP_001288210.1 | 5 | 481 | ||
NM_009642.4 | NP_033772.2 | 5 | 528 | |||
GenBank mRNA | AF102548.1 | - | 5 | 407 | ||
AK007383.1 | - | 5 | 465 | |||
AK007502.1 | - | 5 | 440 | |||
AK036598.1 | - | 5 | 457 | |||
AK152467.1 | - | 5 | 528 | |||
BC046820.1 | - | 5 | 514 | 6400135 | ||
BC057196.1 | - | 5 | 433 | 5249164 |