Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aire | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271549.1 | NP_001258478.1 | 6 | 841 | ||
NM_001271550.1 | NP_001258479.1 | 6 | 841 | |||
NM_001271551.1 | NP_001258480.1 | 6 | 841 | |||
NM_001271552.1 | NP_001258481.1 | 6 | 841 | |||
NM_001271553.1 | NP_001258482.1 | 6 | 841 | |||
NM_001271554.1 | NP_001258483.1 | 6 | 841 | |||
NM_001271555.1 | NP_001258484.1 | 6 | 841 | |||
NM_001271556.1 | NP_001258485.1 | 6 | 841 | |||
NM_001271557.1 | NP_001258486.1 | 6 | 841 | |||
NM_001271558.1 | NP_001258487.1 | 6 | 841 | |||
NM_001271559.1 | NP_001258488.1 | 6 | 841 | |||
NM_009646.2 | NP_033776.1 | 6 | 841 | |||
GenBank mRNA | AF079536.1 | - | 6 | 811 | ||
AF128115.1 | - | 6 | 811 | |||
AF128116.1 | - | 6 | 811 | |||
AF128117.1 | - | 6 | 811 | |||
AF128118.1 | - | 6 | 811 | |||
AF128119.1 | - | 6 | 811 | |||
AF128120.1 | - | 6 | 811 | |||
AF128121.1 | - | 6 | 811 | |||
AF128122.1 | - | 6 | 811 | |||
AF128123.1 | - | 6 | 811 | |||
AF128124.1 | - | 6 | 811 | |||
AF128125.1 | - | 6 | 811 | |||
AF128772.1 | - | 6 | 754 | |||
AF128773.1 | - | 6 | 754 | |||
AJ132243.1 | - | 6 | 754 | |||
AJ243821.1 | - | 6 | 754 | |||
AK153690.1 | - | 7 | 895 | |||
BC103510.1 | - | 6 | 803 | 40039055 | ||
BC103511.1 | - | 6 | 803 | 40039054 | ||
BC103518.1 | - | 6 | 803 | 40039058 | ||
EU625343.3 | - | 6 | 771 | |||
JF267399.1 | - | 6 | 771 |