Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033966.4 | NP_001029138.1 | 6 | 652 | ||
NM_016895.4 | NP_058591.2 | 6 | 652 | |||
GenBank mRNA | AB020202.1 | - | 6 | 574 | ||
AK010936.1 | - | 6 | 571 | |||
AK010951.1 | - | 6 | 573 | |||
AK050133.1 | - | 6 | 613 | |||
AK077519.1 | - | 6 | 576 | |||
AK166976.1 | - | 6 | 563 | |||
AK168056.1 | - | 6 | 575 | |||
AK168148.1 | - | 6 | 602 | |||
BC008610.1 | - | 6 | 562 | 3584786 |