Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005077.1 | - | 2 | 510 | ||
AK039267.1 | - | 1 | 170 | |||
BC004802.1 | - | 3 | 341 | 3589325 | ||
BC008184.1 | - | 3 | 341 | 3589205 | ||
S72537.1 | - | 3 | 365 | |||
X03796.1 | - | 3 | 365 |