Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110794.1 | NP_001104264.1 | 12, 13 | 1517 | ||
NM_009674.3 | NP_033804.2 | 12, 13 | 1632 | |||
GenBank mRNA | AK077579.1 | - | 12, 13 | 1325 | ||
AK134589.1 | - | 13, 14 | 1402 | |||
AK149654.1 | - | 12, 13 | 1323 | |||
AK151438.1 | - | 12, 13 | 1326 | |||
AK161613.1 | - | 12, 13 | 1326 | |||
AK167588.1 | - | 12, 13 | 1300 | |||
AK169199.1 | - | 12, 13 | 1299 | |||
AK178718.1 | - | 1, 2 | 104 | |||
BC008997.1 | - | 12, 13 | 1316 | 3157926 | ||
L13129.1 | - | 12, 13 | 1319 |