Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301211.1 | NP_001288140.1 | 7, 8 | 1129 | ||
NM_009677.6 | NP_033807.2 | 8, 9 | 1138 | |||
GenBank mRNA | AK034198.1 | - | 5, 6 | 367 | ||
AK036387.1 | - | 8, 9 | 1081 | |||
AK145821.1 | - | 7, 8 | 1060 | |||
BC006691.1 | - | 7, 8 | 805 | |||
BC052703.1 | - | 8, 9 | 1011 | 5698075 | ||
BC054535.1 | - | 8, 9 | 1011 | 5698075 | ||
X54424.1 | - | 7, 8 | 1051 |