Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF068707.1 | - | 7 | 797 | ||
AK052384.1 | - | 7 | 732 | |||
AK147177.1 | - | 8 | 836 | |||
AK151844.1 | - | 8 | 828 | |||
BC058402.1 | - | 6 | 842 | |||
BC080798.1 | - | 7 | 745 | |||
BC108374.1 | - | 8 | 823 | 4936738 |